Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014113.2 | NP_001014135.2 | 10 - 11 | 992 | 76 | |
GenBank mRNA | BC099799.1 | - | 10 - 11 | 992 | 7936638 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |