Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173429.1 | NP_001166900.1 | 8 - 9 | 1144 | 69 | |
XM_006257463.2 | XP_006257525.1 | 8 - 9 | 1467 | 69 | ||
XM_008773441.1 | XP_008771663.1 | 8 - 9 | 1467 | 69 | ||
XM_008773442.1 | XP_008771664.1 | 8 - 9 | 1467 | 69 | ||
XM_008773443.1 | XP_008771665.1 | 8 - 9 | 1467 | 69 | ||
XM_008773444.1 | XP_008771666.1 | 8 - 9 | 1174 | 69 | ||
GenBank mRNA | BC169051.1 | - | 8 - 9 | 1144 | 7381285 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |