Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009419.1 | NP_001009419.1 | 4 - 5 | 689 | 113 | |
GenBank mRNA | BC087599.1 | - | 4 - 5 | 689 | 7313414 | 113 |
FQ230721.1 | - | 3 - 4 | 304 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |