Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172091.2 | NP_742088.1 | 7 - 8 | 796 | 102 | |
NM_172092.2 | NP_742089.1 | 7 - 8 | 804 | 102 | ||
NR_073147.1 | 6 - 7 | 697 | 102 | |||
XM_006247876.2 | XP_006247938.1 | 7 - 8 | 910 | 102 | ||
XM_006247877.1 | XP_006247939.1 | 5 - 6 | 635 | 102 | ||
XM_008768468.1 | XP_008766690.1 | 6 - 7 | 853 | 102 | ||
XM_008768469.1 | XP_008766691.1 | 6 - 7 | 856 | 102 | ||
GenBank mRNA | BC081700.1 | - | 6 - 7 | 661 | 7192550 | 102 |
CV117246.1 | - | 3 - 4 | 320 | 102 | ||
L04796.1 | - | 7 - 8 | 766 | 102 | ||
M96674.1 | - | 7 - 8 | 806 | 102 | ||
X68692.1 | - | 7 - 8 | 766 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |