Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024966.1 | NP_001020137.1 | 5 - 6 | 693 | 85 | |
GenBank mRNA | BC091329.1 | - | 5 - 6 | 693 | 7313977 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |