Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246817.2 | XP_006246879.1 | 5 - 6 | 914 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |