Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727774.2 | XP_002727820.1 | 3 - 4 | 456 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |