Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247568.2 | XP_006247630.1 | 1 - 2 | 89 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |