Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107281.1 | NP_001100751.1 | 3 - 4 | 294 | 113 | |
XM_006252408.2 | XP_006252470.1 | 3 - 4 | 296 | 113 | ||
XM_006252409.2 | XP_006252471.1 | 3 - 4 | 291 | 113 | ||
XM_006252410.2 | XP_006252472.1 | 3 - 4 | 362 | 113 | ||
XM_006252413.2 | XP_006252475.1 | 2 - 3 | 144 | 113 | ||
XM_008770870.1 | XP_008769092.1 | 2 - 3 | 2098 | 113 | ||
XM_008770871.1 | XP_008769093.1 | 4 - 5 | 308 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |