Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012012.2 | NP_001012012.2 | 3 - 4 | 202 | 62 | |
NM_001199169.1 | NP_001186098.1 | 2 - 3 | 142 | 62 | ||
XM_006247195.2 | XP_006247257.1 | 4 - 5 | 595 | 62 | ||
XM_006247196.2 | XP_006247258.1 | 3 - 4 | 372 | 62 | ||
XM_006247198.2 | XP_006247260.1 | 2 - 3 | 312 | 62 | ||
XM_008768047.1 | XP_008766269.1 | 4 - 5 | 946 | 62 | ||
XM_008768048.1 | XP_008766270.1 | 4 - 5 | 1096 | 62 | ||
GenBank mRNA | BC086543.1 | - | 3 - 4 | 202 | 7132863 | 62 |
DN932564.1 | - | 137 | 62 | |||
FQ212083.1 | - | 3 - 4 | 173 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |