Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013975.2 | NP_001013997.2 | 4 - 5 | 581 | 99 | |
XM_006247887.2 | XP_006247949.1 | 4 - 5 | 814 | 99 | ||
XM_006247888.2 | XP_006247950.1 | 3 - 4 | 1151 | 99 | ||
GenBank mRNA | EX493402.1 | - | 3 - 4 | 385 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |