Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013103.1 | NP_037235.1 | 7 - 8 | 1371 | 78 | |
GenBank mRNA | BC081826.1 | - | 6 - 7 | 1486 | 7109422 | 78 |
EF121973.1 | - | 7 - 8 | 1344 | 78 | ||
EF121974.1 | - | 7 - 8 | 1266 | 78 | ||
EF122006.1 | - | 7 - 8 | 1344 | 78 | ||
X56546.1 | - | 7 - 8 | 1371 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |