Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105789.1 | NP_001099259.1 | 6 - 7 | 1360 | 82 | |
XM_008767788.1 | XP_008766010.1 | 6 - 7 | 2654 | 82 | ||
XM_008767789.1 | XP_008766011.1 | 6 - 7 | 3214 | 82 | ||
XM_008767790.1 | XP_008766012.1 | 5 - 6 | 1543 | 82 | ||
XM_008767791.1 | XP_008766013.1 | 5 - 6 | 1544 | 82 | ||
XM_008767792.1 | XP_008766014.1 | 5 - 6 | 1542 | 82 | ||
XM_008767793.1 | XP_008766015.1 | 5 - 6 | 1545 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |