Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022296.2 | NP_071632.2 | 2 - 3 | 630 | 87 | |
GenBank mRNA | AJ295749.2 | - | 2 - 3 | 629 | 87 | |
BC105767.1 | - | 2 - 3 | 630 | 7930580 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |