Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252286.2 | XP_006252348.1 | 6 - 7 | 1564 | 101 | |
XM_006252289.1 | XP_006252351.1 | 6 - 7 | 1353 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |