Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025279.1 | NP_001020450.1 | 6 - 7 | 572 | 128 | |
GenBank mRNA | BC097957.1 | - | 6 - 7 | 572 | 7462731 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |