Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764641.1 | XP_008762863.1 | 13 - 14 | 2107 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |