Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100756.1 | NP_001094226.1 | 12 - 13 | 1764 | 69 | |
XM_008764464.1 | XP_008762686.1 | 5 - 6 | 2833 | 69 | ||
GenBank mRNA | BC168226.1 | - | 12 - 13 | 1764 | 7371309 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |