Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106735.1 | NP_001100205.1 | 3 - 4 | 1473 | 67 | |
XM_006240135.2 | XP_006240197.1 | 3 - 4 | 2404 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |