Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007613.1 | NP_001007614.1 | 10 - 11 | 1323 | 145 | |
XM_008768312.1 | XP_008766534.1 | 9 - 10 | 1312 | 145 | ||
XM_008768313.1 | XP_008766535.1 | 9 - 10 | 1235 | 145 | ||
GenBank mRNA | BC079036.1 | - | 10 - 11 | 1323 | 7114313 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |