Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261387.1 | NP_001248316.1 | 6 - 7 | 631 | 140 | |
NM_001261388.1 | NP_001248317.1 | 5 - 6 | 598 | 140 | ||
NM_001261389.1 | NP_001248318.1 | 6 - 7 | 467 | 140 | ||
NM_022929.2 | NP_075218.2 | 5 - 6 | 434 | 140 | ||
XM_006246105.2 | XP_006246167.1 | 5 - 6 | 1489 | 140 | ||
GenBank mRNA | AY082657.1 | - | 6 - 7 | 467 | 140 | |
AY082658.1 | - | 5 - 6 | 434 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |