Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113497.1 | NP_001106969.1 | 382 | 97 | ||
NM_031837.2 | NP_114025.2 | 1270 | 97 | |||
NM_176856.2 | NP_789826.2 | 1389 | 97 | |||
GenBank mRNA | AF170253.1 | - | 1390 | 97 | ||
AF173899.1 | - | 1269 | 97 | |||
AF180525.1 | - | 3 - 4 | 549 | 97 | ||
AF180526.1 | - | 370 | 97 | |||
BC166602.1 | - | 935 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |