Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017477.1 | NP_001017477.1 | 2 - 3 | 489 | 127 | |
XM_006246783.2 | XP_006246845.1 | 3 - 4 | 439 | 127 | ||
XM_006246785.1 | XP_006246847.1 | 3 - 4 | 439 | 127 | ||
XM_008767855.1 | XP_008766077.1 | 3 - 4 | 439 | 127 | ||
XM_008767856.1 | XP_008766078.1 | 3 - 4 | 439 | 127 | ||
GenBank mRNA | BC083699.1 | - | 2 - 3 | 489 | 7190067 | 127 |
FQ233878.1 | - | 3 - 4 | 322 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |