Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtcap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109674.1 | NP_001103144.1 | 2 - 3 | 333 | 72 | |
GenBank mRNA | BC100633.1 | - | 2 - 3 | 280 | 5622000 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |