Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108024.1 | NP_001101494.1 | 3 - 4 | 1860 | 140 | |
XM_008764687.1 | XP_008762909.1 | 1 - 2 | 1460 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |