Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024800.1 | NP_001019971.1 | 4 - 5 | 473 | 144 | |
XM_006240178.2 | XP_006240240.1 | 4 - 5 | 1696 | 144 | ||
GenBank mRNA | BC094308.1 | - | 4 - 5 | 473 | 7316392 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |