Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108702.2 | NP_001102172.2 | 7 - 8 | 1070 | 88 | |
XM_008764486.1 | XP_008762708.1 | 8 - 9 | 1446 | 88 | ||
XM_008764487.1 | XP_008762709.1 | 8 - 9 | 1443 | 88 | ||
XM_008764488.1 | XP_008762710.1 | 7 - 8 | 1351 | 88 | ||
GenBank mRNA | BC166846.1 | - | 7 - 8 | 1070 | 9030077 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |