Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109615.1 | NP_001103085.1 | 4 - 5 | 887 | 58 | |
GenBank mRNA | BC131849.1 | - | 4 - 5 | 844 | 7108272 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |