Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pih1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024868.1 | NP_001020039.1 | 5 - 6 | 813 | 96 | |
XM_006229027.1 | XP_006229089.1 | 6 - 7 | 550 | 96 | ||
XM_006229028.2 | XP_006229090.1 | 6 - 7 | 786 | 96 | ||
GenBank mRNA | BC097946.1 | - | 5 - 6 | 813 | 7366310 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |