Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229263.2 | XP_006229325.1 | 11 - 12 | 2869 | 105 | |
GenBank mRNA | BC099080.1 | - | 6 - 7 | 1218 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |