Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759566.1 | XP_008757788.1 | 9 - 10 | 1605 | 119 | |
XM_008759567.1 | XP_008757789.1 | 9 - 10 | 1886 | 119 | ||
XM_008759568.1 | XP_008757790.1 | 9 - 10 | 1602 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |