Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014161.1 | NP_001014183.1 | 5 - 6 | 709 | 94 | |
GenBank mRNA | BC076398.1 | - | 5 - 6 | 709 | 7110062 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |