Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB195309.1 | - | 14 - 15 | 3008 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |