Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB195309.1 | - | 3 - 4 | 808 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |