Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181082.2 | NP_851596.1 | 2 - 3 | 391 | 113 | |
GenBank mRNA | AY258290.1 | - | 2 - 3 | 391 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |