Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127542.1 | NP_001121014.1 | 1 - 2 | 138 | 63 | |
GenBank mRNA | BC161922.1 | - | 1 - 2 | 124 | 8370772 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |