Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199090.2 | NP_954521.1 | 5 - 6 | 889 | 83 | |
XM_006230760.2 | XP_006230822.1 | 4 - 5 | 791 | 83 | ||
GenBank mRNA | BC060593.1 | - | 5 - 6 | 890 | 6887766 | 83 |
FQ212766.1 | - | 5 - 6 | 836 | 83 | ||
FQ215165.1 | - | 5 - 6 | 817 | 83 | ||
FQ219356.1 | - | 5 - 6 | 840 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |