Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106333.1 | NP_001099803.1 | 6 - 7 | 1029 | 71 | |
XM_006230763.2 | XP_006230825.1 | 7 - 8 | 1596 | 71 | ||
XM_006230764.2 | XP_006230826.1 | 7 - 8 | 859 | 71 | ||
XM_008760110.1 | XP_008758332.1 | 7 - 8 | 1808 | 71 | ||
XM_008760111.1 | XP_008758333.1 | 7 - 8 | 1792 | 71 | ||
XM_008760112.1 | XP_008758334.1 | 6 - 7 | 958 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |