Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759423.1 | XP_008757645.1 | 3 - 4 | 889 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |