Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207591.1 | NP_997474.1 | 5 - 6 | 669 | 91 | |
GenBank mRNA | AY535002.1 | - | 5 - 6 | 669 | 91 | |
BC086359.1 | - | 5 - 6 | 673 | 7304013 | 91 | |
FQ212888.1 | - | 5 - 6 | 700 | 91 | ||
FQ213201.1 | - | 5 - 6 | 678 | 91 | ||
FQ215499.1 | - | 3 - 4 | 370 | 91 | ||
FQ218937.1 | - | 5 - 6 | 675 | 91 | ||
FQ219058.1 | - | 5 - 6 | 675 | 91 | ||
FQ222789.1 | - | 5 - 6 | 679 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |