Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080585.1 | NP_542152.1 | 5 - 6 | 465 | 99 | |
GenBank mRNA | BC063156.1 | - | 5 - 6 | 524 | 6921187 | 99 |
FQ213217.1 | - | 5 - 6 | 469 | 99 | ||
FQ216375.1 | - | 5 - 6 | 783 | 99 | ||
X89968.1 | - | 5 - 6 | 465 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |