Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249579.2 | XP_006249641.1 | 2 - 3 | 256 | 69 | |
XM_213799.8 | XP_213799.6 | 2 - 3 | 256 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |