Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271194.1 | NP_001258123.1 | 1416 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |