Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033062.1 | NP_001028234.1 | 1 | 1570 | 67 | |
GenBank mRNA | BC100630.1 | - | 1 | 1570 | 7938080 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |