Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031813.2 | NP_001026983.1 | 4 - 5 | 492 | 72 | |
NM_001170477.1 | NP_001163948.1 | 5 - 6 | 654 | 72 | ||
GenBank mRNA | BC097985.1 | - | 4 - 5 | 446 | 7444444 | 72 |
CB315310.1 | - | 4 - 5 | 495 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |