Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134747.1 | NP_001128219.1 | 3 - 4 | 326 | 105 | |
GenBank mRNA | BC168220.1 | - | 3 - 4 | 326 | 7320571 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |