Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025126.1 | NP_001020297.1 | 7 - 8 | 902 | 79 | |
XM_006250113.2 | XP_006250175.1 | 14 - 15 | 2101 | 79 | ||
XM_006250114.2 | XP_006250176.1 | 14 - 15 | 2089 | 79 | ||
XM_006250115.2 | XP_006250177.1 | 13 - 14 | 2041 | 79 | ||
XM_006250116.2 | XP_006250178.1 | 13 - 14 | 2029 | 79 | ||
GenBank mRNA | BC091284.1 | - | 7 - 8 | 902 | 7304516 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |