Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249748.2 | XP_006249810.1 | 9 - 10 | 1438 | 70 | |
XM_006249749.2 | XP_006249811.1 | 10 - 11 | 1496 | 70 | ||
GenBank mRNA | BC062002.1 | - | 9 - 10 | 1243 | 5599861 | 70 |
D45187.1 | - | 9 - 10 | 1375 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |