Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106513.1 | NP_001099983.1 | 1 - 2 | 280 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |