Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271283.1 | NP_001258212.1 | 5 - 6 | 964 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |